Merck
  • Expression of gap junction protein connexin36 in multiple subtypes of GABAergic neurons in adult rat somatosensory cortex.
Expression of gap junction protein connexin36 in multiple subtypes of GABAergic neurons in adult rat somatosensory cortex.
Cerebral cortex (New York, N.Y. : 1991) (2011-04-07)
Yunfei Ma, Hiroyuki Hioki, Michiteru Konno, Shixiu Pan, Hisashi Nakamura, Kouichi C Nakamura, Takahiro Furuta, Jin-Lian Li, Takeshi Kaneko
ABSTRACT
To characterize connexin36 (Cx36)-expressing neurons of the adult rat somatosensory cortex, we examined fluorescence signals for Cx36 messenger RNA (mRNA) in 3 nonoverlapping subpopulations of γ-aminobutyric acid (GABA)ergic interneurons, which showed immunoreactivity for 1) parvalbumin (PV); 2) somatostatin (SOM); and 3) either calretinin (CR), vasoactive intestinal polypeptide (VIP), cholecystokinin (CCK), or choline acetyltransferase (ChAT). About 80% of PV-, 52% of SOM-, 37% of CR/VIP/CCK/ChAT-immunoreactive cells displayed Cx36 signals across all cortical layers, and inversely 64%, 25%, and 9% of Cx36-expressing neurons were positive for PV, SOM, or CR/VIP/CCK/ChAT, respectively. Notably, although almost all Cx36-expressing neurons in layer (L) 4, L5, and L6 were positive for one of these markers, a substantial proportion of those in L1 (91%) and L2/3 (10%) were negative for the markers tested, suggesting that other types of neurons might express Cx36. We further investigated the colocalization of Cx36 mRNA and α-actinin2 immunoreactivity, as a marker for late-spiking GABAergic neurons, by using mirror-image sections. Surprisingly, more than 77% of α-actinin2-positive cells displayed Cx36 signals in L1-L3, and about 49% and 13% of Cx36-expressing neurons were positive for α-actinin2 in L1 and L2/3, respectively. These findings suggest that all the subtypes of GABAergic interneurons might form gap junctions in the neocortex.