Merck
  • GZ-11608, a Vesicular Monoamine Transporter-2 Inhibitor, Decreases the Neurochemical and Behavioral Effects of Methamphetamine.
GZ-11608, a Vesicular Monoamine Transporter-2 Inhibitor, Decreases the Neurochemical and Behavioral Effects of Methamphetamine.
The Journal of pharmacology and experimental therapeutics (2019-08-16)
Na-Ra Lee, Guangrong Zheng, Markos Leggas, Venumadhav Janganati, Justin R Nickell, Peter A Crooks, Michael T Bardo, Linda P Dwoskin
ABSTRACT
Despite escalating methamphetamine use and high relapse rates, pharmacotherapeutics for methamphetamine use disorders are not available. Our iterative drug discovery program had found that R-N-(1,2-dihydroxypropyl)-2,6-cis-di-(4-methoxyphenethyl)piperidine hydrochloride (GZ-793A), a selective vesicular monoamine transporter-2 (VMAT2) inhibitor, specifically decreased methamphetamine's behavioral effects. However, GZ-793A inhibited human-ether-a-go-go-related gene (hERG) channels, suggesting cardiotoxicity and prohibiting clinical development. The current study determined if replacement of GZ-793A's piperidine ring with a phenylalkyl group to yield S-3-(4-methoxyphenyl)-N-(1-phenylpropan-2-yl)propan-1-amine (GZ-11608) diminished hERG interaction while retaining pharmacological efficacy. VMAT2 inhibition, target selectivity, and mechanism of GZ-11608-induced inhibition of methamphetamine-evoked vesicular dopamine release were determined. We used GZ-11608 doses that decreased methamphetamine-sensitized activity to evaluate the potential exacerbation of methamphetamine-induced dopaminergic neurotoxicity. GZ-11608-induced decreases in methamphetamine reinforcement and abuse liability were determined using self-administration, reinstatement, and substitution assays. Results show that GZ-11608 exhibited high affinity (Ki = 25 nM) and selectivity (92-1180-fold) for VMAT2 over nicotinic receptors, dopamine transporter, and hERG, suggesting low side-effects. GZ-11608 (EC50 = 620 nM) released vesicular dopamine 25-fold less potently than it inhibited VMAT2 dopamine uptake. GZ-11608 competitively inhibited methamphetamine-evoked vesicular dopamine release (Schild regression slope = 0.9 ± 0.13). GZ-11608 decreased methamphetamine sensitization without altering striatal dopamine content or exacerbating methamphetamine-induced dopamine depletion, revealing efficacy without neurotoxicity. GZ-11608 exhibited linear pharmacokinetics and rapid brain penetration. GZ-11608 decreased methamphetamine self-administration, and this effect was not surmounted by increasing methamphetamine unit doses. GZ-11608 reduced cue- and methamphetamine-induced reinstatement, suggesting potential to prevent relapse. GZ-11608 neither served as a reinforcer nor substituted for methamphetamine, suggesting low abuse liability. Thus, GZ-11608, a potent and selective VMAT2 inhibitor, shows promise as a therapeutic for methamphetamine use disorder. SIGNIFICANCE STATEMENT: GZ-11608 is a potent and selective vesicular monoamine transporter-2 inhibitor that decreases methamphetamine-induced dopamine release from isolated synaptic vesicles from brain dopaminergic neurons. Results from behavioral studies show that GZ-11608 specifically decreases methamphetamine-sensitized locomotor activity, methamphetamine self-administration, and reinstatement of methamphetamine-seeking behavior, without exhibiting abuse liability. Tolerance does not develop to the efficacy of GZ-11608 to decrease the behavioral effects of methamphetamine. In conclusion, GZ-11608 is an outstanding lead in our search for a therapeutic to treat methamphetamine use disorder.
Sigma-Aldrich
GZ-11608, ≥98% (HPLC)
Supelco
Serotonin, analytical standard