Merck
  • An inhalable nanoparticulate STING agonist synergizes with radiotherapy to confer long-term control of lung metastases.
An inhalable nanoparticulate STING agonist synergizes with radiotherapy to confer long-term control of lung metastases.
Nature communications (2019-11-11)
Yang Liu, William N Crowe, Lulu Wang, Yong Lu, W Jeffrey Petty, Amyn A Habib, Dawen Zhao
ABSTRACT
Mounting evidence suggests that the tumor microenvironment is profoundly immunosuppressive. Thus, mitigating tumor immunosuppression is crucial for inducing sustained antitumor immunity. Whereas previous studies involved intratumoral injection, we report here an inhalable nanoparticle-immunotherapy system targeting pulmonary antigen presenting cells (APCs) to enhance anticancer immunity against lung metastases. Inhalation of phosphatidylserine coated liposome loaded with STING agonist cyclic guanosine monophosphate-adenosine monophosphate (NP-cGAMP) in mouse models of lung metastases enables rapid distribution of NP-cGAMP to both lungs and subsequent uptake by APCs without causing immunopathology. NP-cGAMP designed for enhanced cytosolic release of cGAMP stimulates STING signaling and type I interferons production in APCs, resulting in the pro-inflammatory tumor microenvironment in multifocal lung metastases. Furthermore, fractionated radiation delivered to one tumor-bearing lung synergizes with inhaled NP-cGAMP, eliciting systemic anticancer immunity, controlling metastases in both lungs, and conferring long-term survival in mice with lung metastases and with repeated tumor challenge.
Product Description
Avanti
18:1 Liss Rhod PE, Avanti Research - A Croda Brand
Avanti
18:1 Liss Rhod PE, Avanti Research - A Croda Brand
Avanti
18:0 PA, Avanti Research - A Croda Brand
Sigma-Aldrich
Anti-Luciferase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution