Merck
  • Nanopesticide based on botanical insecticide pyrethrum and its potential effects on honeybees.
Nanopesticide based on botanical insecticide pyrethrum and its potential effects on honeybees.
Chemosphere (2019-07-20)
Cristiane R Oliveira, Caio E C Domingues, Nathalie F S de Melo, Thaisa C Roat, Osmar Malaspina, Monica Jones-Costa, Elaine C M Silva-Zacarin, Leonardo F Fraceto
ABSTRACT
Nanotechnology has the potential to overcome the challenges of sustainable agriculture, and nanopesticides can control agricultural pests and increase farm productivity with little environmental impact. However, it is important to evaluate their toxicity on non-target organisms, such as honeybees (Apis mellifera) that forage on crops. The aims of this study were to develop a nanopesticide that was based on solid lipid nanoparticles (SLNs) loaded with pyrethrum extract (PYR) and evaluate its physicochemical properties and short-term toxicity on a non-target organism (honeybee). SLN + PYR was physicochemically stable after 120 days. SLN + PYR had a final diameter of 260.8 ± 3.7 nm and a polydispersion index of 0.15 ± 0.02 nm, in comparison with SLN alone that had a diameter of 406.7 ± 6.7 nm and a polydispersion index of 0.39 ± 0.12 nm. SLN + PYR had an encapsulation efficiency of 99%. The survival analysis of honeybees indicated that PYR10ng presented shorter longevity than those in the control group (P ≤ 0.01). Empty nanoparticles and PYR10ng caused morphological alterations in the bees' midguts, whereas pyrethrum-loaded nanoparticles had no significant effect on digestive cells, so are considered safer, at least in the short term, for honeybees. These results are important in understanding the effects of nanopesticides on beneficial insects and may decrease the environmental impacts of pesticides.
Product Description
Tripalmitin, NIST® SRM® 1595
Sigma-Aldrich
Poly(L-lactide-co-glycolide), lactide:glycolide 20:80, viscosity 1.6 dL/g