Merck
  • Genome Engineering of Eubacterium limosum Using Expanded Genetic Tools and the CRISPR-Cas9 System.
Genome Engineering of Eubacterium limosum Using Expanded Genetic Tools and the CRISPR-Cas9 System.
ACS synthetic biology (2019-08-03)
Jongoh Shin, Seulgi Kang, Yoseb Song, Sangrak Jin, Jin Soo Lee, Jung-Kul Lee, Dong Rip Kim, Sun Chang Kim, Suhyung Cho, Byung-Kwan Cho
ABSTRACT
Eubacterium limosum is one of the important bacteria in C1 feedstock utilization as well as in human gut microbiota. Although E. limosum has recently garnered much attention and investigation on a genome-wide scale, a bottleneck for systematic engineering in E. limosum is the lack of available genetic tools and an efficient genome editing platform. To overcome this limitation, we here report expanded genetic tools and the CRISPR-Cas9 system. We have developed an inducible promoter system that enables implementation of the CRISPR-Cas9 system to precisely manipulate target genes of the Wood-Ljungdahl pathway with 100% efficiency. Furthermore, we exploited the effectiveness of CRISPR interference to reduce the expression of target genes, exhibiting substantial repression of several genes in the Wood-Ljungdahl pathway and fructose-PTS system. These expanded genetic tools and CRISPR-Cas9 system comprise powerful and widely applicable genetic tools to accelerate functional genomic study and genome engineering in E. limosum.
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for use with bacterial cell extracts, lyophilized powder
Sigma-Aldrich
TargeTron Vector pJIR750ai, Expression Vector for Bacterial Gene Knockout