Merck
  • Functional interactions between nitrite reductase and nitric oxide reductase from Paracoccus denitrificans.
Functional interactions between nitrite reductase and nitric oxide reductase from Paracoccus denitrificans.
Scientific reports (2019-11-23)
Ingrid Albertsson, Johannes Sjöholm, Josy Ter Beek, Nicholas J Watmough, Jerker Widengren, Pia Ädelroth
ABSTRACT
Denitrification is a microbial pathway that constitutes an important part of the nitrogen cycle on earth. Denitrifying organisms use nitrate as a terminal electron acceptor and reduce it stepwise to nitrogen gas, a process that produces the toxic nitric oxide (NO) molecule as an intermediate. In this work, we have investigated the possible functional interaction between the enzyme that produces NO; the cd1 nitrite reductase (cd1NiR) and the enzyme that reduces NO; the c-type nitric oxide reductase (cNOR), from the model soil bacterium P. denitrificans. Such an interaction was observed previously between purified components from P. aeruginosa and could help channeling the NO (directly from the site of formation to the side of reduction), in order to protect the cell from this toxic intermediate. We find that electron donation to cNOR is inhibited in the presence of cd1NiR, presumably because cd1NiR binds cNOR at the same location as the electron donor. We further find that the presence of cNOR influences the dimerization of cd1NiR. Overall, although we find no evidence for a high-affinity, constant interaction between the two enzymes, our data supports transient interactions between cd1NiR and cNOR that influence enzymatic properties of cNOR and oligomerization properties of cd1NiR. We speculate that this could be of particular importance in vivo during metabolic switches between aerobic and denitrifying conditions.
Product Description
Avanti
18:1 Biotinyl PE, Avanti Research - A Croda Brand 870282C
Pricing and availability is not currently available.
Avanti
18:1 Biotinyl PE, Avanti Research - A Croda Brand 870282P, powder
Pricing and availability is not currently available.