Merck
  • Size matters: molecular weight affects the efficiency of poly(ethylenimine) as a gene delivery vehicle.
Size matters: molecular weight affects the efficiency of poly(ethylenimine) as a gene delivery vehicle.
Journal of biomedical materials research (1999-07-09)
W T Godbey, K K Wu, A G Mikos
ABSTRACT
Poly(ethylenimine) (PEI) samples of various molecular weights and pHs were used to transfect endothelial cells to achieve levels of gene expression for comparison. PEIs with nominal molecular weights of 600, 1200, 1800, 10,000, and 70,000 Da were examined at pHs of 5. 0, 6.0, 7.0, and 8.0, and the results were recorded in terms of transfection efficiencies at 24, 48, 68, 92, and 120 h post-transfection. Trials were performed on the human endothelial cell-derived cell line EA.hy 926. We found that, for the polymers tested, transfection efficiency increased as the molecular weight of PEI increased. Representative values of PEIs at pH 6 and molecular weight 70,000 produced average transfection efficiencies of 25.6 +/- 7.9% (n = 8) at the greatest average expression levels, while PEI of molecular weight 10,000 yielded efficiencies of only 11.4 +/- 1.7% (n = 6). Transfection efficiencies for molecular weight 1,800 PEI were essentially zero, and PEIs of lower molecular weights produced no transfection at all. In contrast, the pH of the PEI solutions had no discernible effect on transfection. Optimal expression of the green fluorescent protein reporter occurred between 2 and 3 days post-transfection. The amount of reporter expression also was noted, as determined by the brightness of fluorescing cells under UV. The data obtained demonstrate that the molecular weight of the PEI carrier has an effect on transfection efficiency while the pH of the PEI solutions prior to DNA complexation has no such effect.
Sigma-Aldrich
Branched polyethylenimine solution, 2mg/mL aqueous solution, suitable for biomedical research
Pricing and availability is not currently available.