Merck
  • ERKs and JNKs mediate hydrogen peroxide-induced Egr-1 expression and nuclear accumulation in H9c2 cells.
ERKs and JNKs mediate hydrogen peroxide-induced Egr-1 expression and nuclear accumulation in H9c2 cells.
Physiological research (2009-08-18)
I K S Aggeli, I Beis, C Gaitanaki
ABSTRACT
One of the most significant insults that jeopardize cardiomyocyte homeostasis is a surge of reactive oxygen species (ROS) in the failing myocardium. Early growth response factor-1 (Egr-1) has been found to act as a transcriptional regulator in multiple biological processes known to exert deleterious effects on cardiomyocytes. We thus investigated the signaling pathways involved in its regulation by H2O2. Egr-1 mRNA levels were found to be maximally induced after 2 h in H2O2-treated H9c2 cells. Egr-1 respective response at the protein level, was found to be maximally induced after 2 h of treatment with 200 microM H2O2, remaining elevated for 6 h, and declining thereafter. H2O2-induced upregulation of Egr-1 mRNA and protein levels was ablated in the presence of agents inhibiting ERKs pathway (PD98059) and JNKs (SP600125, AS601245). Immunofluorescent experiments revealed H2O2-induced Egr-1 nuclear sequestration to be also ERK- and JNK-dependent. Overall, our results show for the first time the fundamental role of ERKs and JNKs in regulating Egr-1 response to H2O2 treatment in cardiac cells at multiple levels: mRNA, protein and subcellular distribution. Nevertheless, further studies are required to elucidate the specific physiological role of Egr-1 regarding the modulation of gene expression and determination of cell fate.