Merck
  • Effects of lipoxin A4 on chemotaxis and degranulation of human eosinophils stimulated by platelet-activating factor and N-formyl-L-methionyl-L-leucyl-L-phenylalanine.
Effects of lipoxin A4 on chemotaxis and degranulation of human eosinophils stimulated by platelet-activating factor and N-formyl-L-methionyl-L-leucyl-L-phenylalanine.
Allergy (1994-04-01)
O Soyombo, B W Spur, T H Lee
ABSTRACT
Lipoxins are trihydroxytetraene metabolites derived through a double lipoxygenation of arachidonic acid. Lipoxin A4 (LXA4) was prepared by total chemical synthesis, and its capacity to modulate eosinophil migration has been evaluated. LXA4 is a weak and partial chemotactic agent; at 10(-6) M, it achieved about 20% of the response of 10(-6) M platelet-activating factor (PAF). Preincubation of eosinophils with increasing doses of LXA4 (10(-10)-10(-5) M) resulted in a concentration-dependent inhibition of cell migration induced by 10(-6) M formyl-methionyl-leucyl-phenylalanine (FMLP) and 10(-6) M PAF. The concentration of LXA4 which produced 50% inhibition (IC50) of eosinophil migration was approximately 10(-6) M. LXA4 (10(-10)-10(-6) M) did not elicit ECP release or modulate ECP release induced by 10(-6) M FMLP. LXA4 may have antiallergic properties in preventing eosinophilic migration.