Merck
  • Protective effects of zingerone on cisplatin-induced nephrotoxicity in female rats.
Protective effects of zingerone on cisplatin-induced nephrotoxicity in female rats.
Environmental science and pollution research international (2019-06-06)
Fatih Mehmet Kandemir, Serkan Yildirim, Cuneyt Caglayan, Sefa Kucukler, Gizem Eser
ABSTRACT
Zingerone (ZO), one of the active components of ginger (Zingiber officinale), is a phenolic alkanone with antioxidant, antiapoptotic, and anti-inflammatory properties. Cisplatin (CP) is a widely used chemotherapeutic drug for solid tumors, but its therapeutic use is limited due to dose-dependent nephrotoxicity. In the present study, we investigated the ameliorative effect of ZO against CP-induced nephrotoxicity. Intraperitoneal administration of single-dose CP (7 mg/kg body weight) on the first day enhanced kidney lipid peroxidation and reduced antioxidant enzyme activities such as catalase (CAT), superoxide dismutase (SOD), glutathione peroxidase (GPx), and glutathione (GSH). CP increased serum urea and creatinine levels and disrupted histological integrity while causing a decrease aquaporin 1 (AQP1) level in the kidney tissues. CP induced inflammatory responses by elevating the levels of tumor necrosis factor-α (TNF-α), interleukin-1β (IL-1β), interleukin-6 (IL-6), interleukin-33 (IL-33) and nuclear factor kappa B (NF-κB), and activities of inducible nitric oxide synthase (iNOS) and cyclooxygenase-2 (COX-2). Moreover, it also caused oxidative DNA damage and activation of apoptotic pathway by increasing of 8-hydroxy-2'-deoxyguanosine (8-OHdG), p53, cysteine aspartate-specific protease-3 (caspase-3), and Bcl-2-associated x protein (bax) while decreasing B cell lymphoma-2 (Bcl-2). However, treatment with ZO at a dose of 25 and 50 mg/kg b.wt. for 7 days significantly decreased oxidative stress, apoptosis, inflammation, and histopathological alterations while increased AQP1 levels in the kidney tissue. The results of the current study suggested that ZO as an effective natural product attenuates CP-induced nephrotoxicity.