Merck
  • Regional and correlative sweat analysis using high-throughput microfluidic sensing patches toward decoding sweat.
Regional and correlative sweat analysis using high-throughput microfluidic sensing patches toward decoding sweat.
Science advances (2019-08-28)
Hnin Yin Yin Nyein, Mallika Bariya, Liisa Kivimäki, Sanna Uusitalo, Tiffany Sun Liaw, Elina Jansson, Christine Heera Ahn, John A Hangasky, Jiangqi Zhao, Yuanjing Lin, Tuomas Happonen, Minghan Chao, Christina Liedert, Yingbo Zhao, Li-Chia Tai, Jussi Hiltunen, Ali Javey
ABSTRACT
Recent technological advancements in wearable sensors have made it easier to detect sweat components, but our limited understanding of sweat restricts its application. A critical bottleneck for temporal and regional sweat analysis is achieving uniform, high-throughput fabrication of sweat sensor components, including microfluidic chip and sensing electrodes. To overcome this challenge, we introduce microfluidic sensing patches mass fabricated via roll-to-roll (R2R) processes. The patch allows sweat capture within a spiral microfluidic for real-time measurement of sweat parameters including [Na+], [K+], [glucose], and sweat rate in exercise and chemically induced sweat. The patch is demonstrated for investigating regional sweat composition, predicting whole-body fluid/electrolyte loss during exercise, uncovering relationships between sweat metrics, and tracking glucose dynamics to explore sweat-to-blood correlations in healthy and diabetic individuals. By enabling a comprehensive sweat analysis, the presented device is a crucial tool for advancing sweat testing beyond the research stage for point-of-care medical and athletic applications.