Merck
  • Identification of candidate diagnostic and prognostic biomarkers for human prostate cancer: RPL22L1 and RPS21.
Identification of candidate diagnostic and prognostic biomarkers for human prostate cancer: RPL22L1 and RPS21.
Medical oncology (Northwood, London, England) (2019-05-16)
Zumu Liang, Qingjie Mou, Zhiwei Pan, Qiang Zhang, Guojun Gao, Yingying Cao, Zhiqin Gao, Zhifang Pan, Weiguo Feng
ABSTRACT
Prostate cancer (PCa) is one of the most common malignancies in men worldwide. This study was designed to investigate the potential of Ribosomal Protein L22-like1 (RPL22L1) and Ribosomal Protein S21 (RPS21) as diagnostic and prognostic biomarkers for PCa. First, RPL22L1 and RPS21 were screened as the key molecular of PCa by bioinformatics analysis. Subsequently, the prostate tissue samples were stained for antibodies against RPL22L1 and RPS21. The unbiased signal quantification was performed by ImageJ software, and the results showed that the expression of RPL22L1 and RPS21 exhibited significant differences between the PCa tissues and the normal prostate tissues. Receiver-operating characteristics (ROC) curves were prepared, and then the area under the curve (AUC) values of RPL22L1 and RPS21 were calculated as 0.798 and 0.768, and the likelihood ratio (LR) values of RPL22L1 and RPS21 were calculated as 2.86 and 2.53. These data implied that the over-expression of RPL22L1 and RPS21 is associated with the presence of PCa. The further analysis suggested that the expression of RPL22L1 and RPS21 were significantly higher in high Gleason grade than they were in low Gleason grade. In addition, in vitro studies were undertaken to evaluate the roles of RPL22L1 and RPS21 in PCa. The results revealed that these genes promote PCa cell proliferation, migration and invasion, and inhibit PCa cell apoptosis. Taken together, these data showed that RPL22L1 and RPS21 exhibited higher expression in human prostate cancer tissue, and involved in PCa cell proliferation and invasion. This research provided a novel insight into diagnostic and prognostic biomarkers for PCa.