Merck
  • A C19MC-LIN28A-MYCN Oncogenic Circuit Driven by Hijacked Super-enhancers Is a Distinct Therapeutic Vulnerability in ETMRs: A Lethal Brain Tumor.
A C19MC-LIN28A-MYCN Oncogenic Circuit Driven by Hijacked Super-enhancers Is a Distinct Therapeutic Vulnerability in ETMRs: A Lethal Brain Tumor.
Cancer cell (2019-07-10)
Patrick Sin-Chan, Iqra Mumal, Tannu Suwal, Ben Ho, Xiaolian Fan, Irtisha Singh, Yuchen Du, Mei Lu, Neilket Patel, Jonathon Torchia, Dean Popovski, Maryam Fouladi, Paul Guilhamon, Jordan R Hansford, Sarah Leary, Lindsey M Hoffman, Jean M Mulcahy Levy, Alvaro Lassaletta, Palma Solano-Paez, Eloy Rivas, Alyssa Reddy, G Yancey Gillespie, Nalin Gupta, Timothy E Van Meter, Hideo Nakamura, Tai-Tong Wong, Young-Shin Ra, Seung-Ki Kim, Luca Massimi, Richard G Grundy, Jason Fangusaro, Donna Johnston, Jennifer Chan, Lucie Lafay-Cousin, Eugene I Hwang, Yin Wang, Daniel Catchpoole, Jean Michaud, Benjamin Ellezam, Ramya Ramanujachar, Holly Lindsay, Michael D Taylor, Cynthia E Hawkins, Eric Bouffet, Nada Jabado, Sheila K Singh, Claudia L Kleinman, Dalia Barsyte-Lovejoy, Xiao-Nan Li, Peter B Dirks, Charles Y Lin, Stephen C Mack, Jeremy N Rich, Annie Huang
ABSTRACT
Embryonal tumors with multilayered rosettes (ETMRs) are highly lethal infant brain cancers with characteristic amplification of Chr19q13.41 miRNA cluster (C19MC) and enrichment of pluripotency factor LIN28A. Here we investigated C19MC oncogenic mechanisms and discovered a C19MC-LIN28A-MYCN circuit fueled by multiple complex regulatory loops including an MYCN core transcriptional network and super-enhancers resulting from long-range MYCN DNA interactions and C19MC gene fusions. Our data show that this powerful oncogenic circuit, which entraps an early neural lineage network, is potently abrogated by bromodomain inhibitor JQ1, leading to ETMR cell death.
Sigma-Aldrich
Anti-RBL2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid