Merck
  • C-terminus of heat shock protein 60 can activate macrophages by lectin-like oxidized low-density lipoprotein receptor 1.
C-terminus of heat shock protein 60 can activate macrophages by lectin-like oxidized low-density lipoprotein receptor 1.
Biochemical and biophysical research communications (2018-12-17)
Baonian Liu, Shaoling Li, Bingqiu Xiu, Yaqi Zhang, Yuxiang Zhou, Qinrui Yang, Wanjun Qi, Weicheng Wu, Lan Wang, Jianxin Gu, Jianhui Xie
ABSTRACT
Immune responses against antigens generally require an efficient activation of antigen-presenting cells (APCs). Currently, the targeting of vaccine antigens to APCs has emerged as a promising strategy for boosting vaccine immunogenicity. Here, we reported that the C-terminus of heat shock protein 60 (HSP60C) can activate mouse peritoneal macrophages to secret a series of cytokines, and phosphorylation of p38 mitogen-activated protein kinase (MAPK) and NF-κB p65 was involved in the pathway. We showed that the activation effect of HSP60C on macrophages was independent of toll-like receptor (TLR) 4 and the TLR-associated myeloide differentiation factor 88 (MyD88). Knockdown of lectin-like oxidized low-density lipoprotein receptor-1 (LOX-1) reduced the activation of HSP60C-induced macrophage p38 MAPK, NF-κB p65 and cytokine secretion to some extent. Finally, we found that HSP60C up-regulated the expression of LOX-1 on macrophages and ovalbumin (OVA) model antigen fused with HSP60C markedly enhanced OVA-specific IgG responses. Thus, our results unravel a novel LOX-1-dependent pathway by which HSP60C can effectively activate macrophages and APCs targeting based on LOX-1 interaction is a promising approach to improve vaccines.