Merck
  • TCF/LEF dependent and independent transcriptional regulation of Wnt/β-catenin target genes.
TCF/LEF dependent and independent transcriptional regulation of Wnt/β-catenin target genes.
The EMBO journal (2018-11-15)
Nikolaos Doumpas, Franziska Lampart, Mark D Robinson, Antonio Lentini, Colm E Nestor, Claudio Cantù, Konrad Basler
ABSTRACT
During canonical Wnt signalling, the activity of nuclear β-catenin is largely mediated by the TCF/LEF family of transcription factors. To challenge this view, we used the CRISPR/Cas9 genome editing approach to generate HEK 293T cell clones lacking all four TCF/LEF genes. By performing unbiased whole transcriptome sequencing analysis, we found that a subset of β-catenin transcriptional targets did not require TCF/LEF factors for their regulation. Consistent with this finding, we observed in a genome-wide analysis that β-catenin occupied specific genomic regions in the absence of TCF/LEF Finally, we revealed the existence of a transcriptional activity of β-catenin that specifically appears when TCF/LEF factors are absent, and refer to this as β-catenin-GHOST response. Collectively, this study uncovers a previously neglected modus operandi of β-catenin that bypasses the TCF/LEF transcription factors.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture