Merck
  • Proteome profiling reveals potential cellular targets of staurosporine using a clickable cell-permeable probe.
Proteome profiling reveals potential cellular targets of staurosporine using a clickable cell-permeable probe.
Chemical communications (Cambridge, England) (2011-09-17)
Haibin Shi, Xiamin Cheng, Siu Kwan Sze, Shao Q Yao
ABSTRACT
A clickable, affinity-based probe (AfBP), which was modified from staurosporine (a natural product kinase inhibitor), has been synthesized and used in situ for activity-based proteome profiling of potential cellular targets of staurosporine in HepG2 cancer cells.
Sigma-Aldrich
Fmoc-L-photo-methionine, ā‰„95%