Merck
Pralatrexate pharmacology and clinical development.
Clinical cancer research : an official journal of the American Association for Cancer Research (2013-08-24)
Enrica Marchi, Michael Mangone, Kelly Zullo, Owen A O'Connor
ABSTRACT
Folates are well known to be essential for many cellular processes, including cellular proliferation. As a consequence, antifolates, the fraudulent mimics of folic acid, have been shown to be potent therapeutic agents in many cancers. Over the past several decades, efforts to improve on this class of drugs have met with little success. Recently, one analog specifically designed to have high affinity for the reduced folate carrier, which efficiently internalizes natural folates and antifolates, has been shown to be very active in T-cell lymphoma. Pralatrexate, approved by the U.S. Food and Drug Administration in 2009, is highly active across many lymphoid malignancies, including chemotherapy-resistant T-cell lymphoma. Emerging combination studies have now shown that pralatrexate is highly synergistic with gemcitabine, histone deacetylase inhibitors like romidepsin and bortezomib. These insights are leading to a number of novel phase I and II combination studies which could challenge existing regimens like CHOP, and improve the outcome of patients with T-cell lymphoma.
Sigma-Aldrich
Pralatrexate, ≥98% (HPLC)