Merck
  • Two contrary functions of tenascin: dissection of the active sites by recombinant tenascin fragments.
Two contrary functions of tenascin: dissection of the active sites by recombinant tenascin fragments.
Cell (1989-10-20)
J Spring, K Beck, R Chiquet-Ehrismann
ABSTRACT
A structural and functional model of tenascin was elaborated using recombinant parts of three alternatively spliced tenascin variants and anti-tenascin monoclonal antibodies. The fusion proteins were compared with intact tenascin for their functions and by electron microscopy. A strong cell binding site was localized within 104 amino acids. This fragment also contains the epitope of the monoclonal antibody anti-Tn68, which inhibits cell attachment to tenascin and binds near the tips of the six arms of tenascin. In contrast, constructs containing the 13 1/2 EGF-like repeats of tenascin showed an antiadhesive effect. The coexistence of the two contrary signals on the same molecule might be responsible for the versatile features of tenascin.