Merck
  • No long-term weight maintenance effects of gelatin in a supra-sustained protein diet.
No long-term weight maintenance effects of gelatin in a supra-sustained protein diet.
Physiology & behavior (2010-05-12)
A Hochstenbach-Waelen, K R Westerterp, S Soenen, M S Westerterp-Plantenga
ABSTRACT
In the short-term, gelatin showed stronger hunger suppression and less energy intake compared with other proteins. This study investigated if a supra-sustained gelatin-milk protein (GMP) diet improves weight maintenance (WM) compared with a sustained milk protein (SMP) diet and supra-sustained milk protein (SSMP) diet during a 4-months WM period after 8-week weight loss (WL) in sixty-five healthy subjects (28.6+/-3.4kg/m(2); 44+/-10years). Absolute protein intake was kept constant (sustained) throughout per subject. Diets were: protein(P)/fat(F)/carbohydrate(C): 15/40/45% of energy (En%) (SMP) and 30/25/45 En% (SSMP or GMP) for weeks 9-16. Diets on weeks 17-24: P/F/C: 30/35/35 En% (SMP) and 60/5/35 En% (SSMP or GMP). From weeks 8 to 16, and weeks 16 to 24, changes in BMI were similar between the GMP (-0.4+/-0.6 and 0.3+/-0.7kg/m(2) respectively), and the SMP (-0.7+/-0.9 and 0.1+/-0.7kg/m(2) respectively) and SSMP (-0.6+/-0.6 and 0.3+/-0.6kg/m(2) respectively) diets. Sparing of fat free mass (FFM): increases/decreases in FFM%/fat-mass% from weeks 8 to 16 were similar between the GMP and both control diets, and maintained from weeks 16 to 24. In conclusion, all 3 diets resulted in a successful WM period, while a GMP diet does not improve body weight maintenance and related variables after weight loss compared with a SMP and SSMP diet.