Merck
  • The role of excitatory amino acids and NMDA receptors in traumatic brain injury.
The role of excitatory amino acids and NMDA receptors in traumatic brain injury.
Science (New York, N.Y.) (1989-05-19)
A I Faden, P Demediuk, S S Panter, R Vink
ABSTRACT
Brain injury induced by fluid percussion in rats caused a marked elevation in extracellular glutamate and aspartate adjacent to the trauma site. This increase in excitatory amino acids was related to the severity of the injury and was associated with a reduction in cellular bioenergetic state and intracellular free magnesium. Treatment with the noncompetitive N-methyl-D-aspartate (NMDA) antagonist dextrophan or the competitive antagonist 3-(2-carboxypiperazin-4-yl)propyl-1-phosphonic acid limited the resultant neurological dysfunction; dextrorphan treatment also improved the bioenergetic state after trauma and increased the intracellular free magnesium. Thus, excitatory amino acids contribute to delayed tissue damage after brain trauma; NMDA antagonists may be of benefit in treating acute head injury.