Merck
  • A Lateral Flow Immunochromato-graphic Strip Test for Rapid Detection of Oseltamivir Phosphate in Egg and Chicken Meat.
A Lateral Flow Immunochromato-graphic Strip Test for Rapid Detection of Oseltamivir Phosphate in Egg and Chicken Meat.
Scientific reports (2018-11-14)
Xingdong Yang, Jifei Yang, Yinbiao Wang, Lili Li, Zhongke Sun, Zonghao Yue, Fengshou Tian, Le He, Xiaofei Hu
ABSTRACT
A lateral flow immunochromatographic strip test (LFIST) based on a competitive format was developed for rapid and sensitive on-site detection of oseltamivir phosphate (OP) residues in poultry product. The sensitivity (half inhibitory concentration, IC50) of the LFIST in the detection of egg and chicken meat samples was confirmed to be 2.56 and 2.63 µg/kg, and the limit detection (LOD) value were 0.43 and 0.42 µg/kg, respectively. For intra-assay and inter-assay reproducibility, recoveries of OP spiked samples ranged between 82.8% and 91.2% with coefficients of variations (CV) less than 5.67% (intra-assay) and 6.52% (inter-assay). The performance of LFIST was comparable to high-performance liquid chromatography (HPLC) in a parallel testing of egg samples and chicken samples. LFIST takes less than 5 minutes, eliminates the dependency on professional personnel, and thus can be used as a surveillance tool for on-site detection of OP residues.
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl 4-bromobutyrate, 95%
Pricing and availability is not currently available.