Merck
  • Thermodynamic and computational analyses reveal the functional roles of the galloyl group of tea catechins in molecular recognition.
Thermodynamic and computational analyses reveal the functional roles of the galloyl group of tea catechins in molecular recognition.
PloS one (2018-10-12)
Tomoya Takahashi, Satoru Nagatoishi, Daisuke Kuroda, Kouhei Tsumoto
ABSTRACT
Catechins, biologically active polyphenols in green tea, exhibit various biological activities, such as anticancer and antiviral activities, arising from interactions with functional proteins. However, the molecular details of these interactions remain unclear. In this study, we investigated the interactions between human serum albumin (HSA) and various catechins, including some with a galloyl group, by means of isothermal titration calorimetry (ITC), differential scanning calorimetry (DSC), and docking simulations. Our results indicate that the galloyl group was important for recognition by HSA and was responsible for enthalpic gains derived from a larger buried surface area and more van der Waals contacts. Thus, our thermodynamic and computational analyses suggest that the galloyl group plays important functional roles in the specific binding of catechins to proteins, implying that the biological activities of these compounds may be due in part to the physicochemical characteristics of the galloyl group.
Sigma-Aldrich
Ethyl gallate, antioxidant, ≥96.0% (HPLC)