Merck
  • Upgrading traditional liquid electrolyte via in situ gelation for future lithium metal batteries.
Upgrading traditional liquid electrolyte via in situ gelation for future lithium metal batteries.
Science advances (2018-10-13)
Feng-Quan Liu, Wen-Peng Wang, Ya-Xia Yin, Shuai-Feng Zhang, Ji-Lei Shi, Lu Wang, Xu-Dong Zhang, Yue Zheng, Jian-Jun Zhou, Lin Li, Yu-Guo Guo
ABSTRACT
High-energy lithium metal batteries (LMBs) are expected to play important roles in the next-generation energy storage systems. However, the uncontrolled Li dendrite growth in liquid electrolytes still impedes LMBs from authentic commercialization. Upgrading the traditional electrolyte system from liquid to solid and quasi-solid has therefore become a key issue for prospective LMBs. From this premise, it is particularly urgent to exploit facile strategies to accomplish this goal. We report that commercialized liquid electrolyte can be easily converted into a novel quasi-solid gel polymer electrolyte (GPE) via a simple and efficient in situ gelation strategy, which, in essence, is to use LiPF6 to induce the cationic polymerization of the ether-based 1,3-dioxolane and 1,2-dimethoxyethane liquid electrolyte under ambient temperature. The newly developed GPE exhibits elevated protective effects on Li anodes and has universality for diversified cathodes including but not restricted to sulfur, olivine-type LiFePO4, and layered LiNi0.6Co0.2Mn0.2O2, revealing tremendous potential in promoting the large-scale application of future LMBs.