Merck
  • EPHB6 augments both development and drug sensitivity of triple-negative breast cancer tumours.
EPHB6 augments both development and drug sensitivity of triple-negative breast cancer tumours.
Oncogene (2018-04-28)
Behzad M Toosi, Amr El Zawily, Luke Truitt, Matthew Shannon, Odette Allonby, Mohan Babu, John DeCoteau, Darrell Mousseau, Mohsin Ali, Tanya Freywald, Amanda Gall, Frederick S Vizeacoumar, Morgan W Kirzinger, C Ronald Geyer, Deborah H Anderson, TaeHyung Kim, Alana L Welm, Peter Siegel, Franco J Vizeacoumar, Anthony Kusalik, Andrew Freywald
ABSTRACT
Triple-negative breast cancer (TNBC) tumours that lack expression of oestrogen, and progesterone receptors, and do not overexpress the HER2 receptor represent the most aggressive breast cancer subtype, which is characterised by the resistance to therapy in frequently relapsing tumours and a high rate of patient mortality. This is likely due to the resistance of slowly proliferating tumour-initiating cells (TICs), and understanding molecular mechanisms that control TICs behaviour is crucial for the development of effective therapeutic approaches. Here, we present our novel findings, indicating that an intrinsically catalytically inactive member of the Eph group of receptor tyrosine kinases, EPHB6, partially suppresses the epithelial-mesenchymal transition in TNBC cells, while also promoting expansion of TICs. Our work reveals that EPHB6 interacts with the GRB2 adapter protein and that its effect on enhancing cell proliferation is mediated by the activation of the RAS-ERK pathway, which allows it to elevate the expression of the TIC-related transcription factor, OCT4. Consistent with this, suppression of either ERK or OCT4 activities blocks EPHB6-induced pro-proliferative responses. In line with its ability to trigger propagation of TICs, EPHB6 accelerates tumour growth, potentiates tumour initiation and increases TIC populations in xenograft models of TNBC. Remarkably, EPHB6 also suppresses tumour drug resistance to DNA-damaging therapy, probably by forcing TICs into a more proliferative, drug-sensitive state. In agreement, patients with higher EPHB6 expression in their tumours have a better chance for recurrence-free survival. These observations describe an entirely new mechanism that governs TNBC and suggest that it may be beneficial to enhance EPHB6 action concurrent with applying a conventional DNA-damaging treatment, as it would decrease drug resistance and improve tumour elimination.