Merck
  • Elucidating the transactivation domain of the pleiotropic transcription factor Myrf.
Elucidating the transactivation domain of the pleiotropic transcription factor Myrf.
Scientific reports (2018-09-01)
Jin-Ok Choi, Chuandong Fan, Dongkyeong Kim, Mohamed Sharif, Hongjoo An, Yungki Park
ABSTRACT
Myrf is a newly discovered membrane-bound transcription factor that plays an essential role in as diverse organisms as human, worm, and slime mold. Myrf is generated as a type-II membrane protein in the endoplasmic reticulum (ER). It forms homo-oligomers to undergo auto-cleavage that releases Myrf N-terminal fragment from the ER membrane as a homo-trimer. The homo-trimer of Myrf N-terminal fragments enters the nucleus and binds the Myrf motif to activate transcription. Despite its prominent role as a transcriptional activator, little is known about the transactivation domain of Myrf. Here, we report that the N-terminal-most (NTM) domain of Myrf is required for transcriptional activity and, when fused to a Gal4 fragment, enables it to activate transcription. The transactivation function of the NTM domain did not require homo-trimerization. We also discovered that the NTM domain can be sumoylated at three lysine residues (K123, K208, and K276), with K276 serving as the main acceptor. K276 sumoylation repressed the transactivation function of the NTM domain without affecting the stability or nuclear localization of Myrf N-terminal fragment. In sum, this study identifies the NTM domain as the transactivation domain of Myrf and the potential regulatory impact of its K276 sumoylation.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Millipore
EZview Red Anti-HA Affinity Gel