Merck
  • Conantokin-T. A gamma-carboxyglutamate containing peptide with N-methyl-d-aspartate antagonist activity.
Conantokin-T. A gamma-carboxyglutamate containing peptide with N-methyl-d-aspartate antagonist activity.
The Journal of biological chemistry (1990-04-15)
J A Haack, J Rivier, T N Parks, E E Mena, L J Cruz, B M Olivera
ABSTRACT
Conantokin-T, a 21-amino acid peptide which induces sleep-like symptoms in young mice was purified from the venom of the fish-hunting cone snail, Conus tulipa. The amino acid sequence of the peptide was determined and verified by chemical synthesis. The peptide has 4 residues of the modified amino acid, gamma-carboxyglutamate (Gla). The sequence of the peptide is: Gly-Glu-Gla-Gla-Tyr-Gln-Lys-Met-Leu-Gla-Asn-Leu-Arg-Gla-Ala-Glu-Val-Lys- Lys-Asn-Ala-NH2. Conantokin-T inhibits N-methyl-D-aspartate (NMDA) receptor-mediated calcium influx in central nervous system neurons. This observation suggests that like conantokin-G (a homologous Conus peptide with recently identified NMDA antagonist activity) conantokin-T has NMDA antagonist activity. A sequence comparison of conantokins-T and -G identifies the 4 Gla residues and the N-terminal dipeptide sequence as potential key elements for the biological activity of this peptide.