Merck
  • Adult-born dentate granule cell excitability depends on the interaction of neuron age, ontogenetic age and experience.
Adult-born dentate granule cell excitability depends on the interaction of neuron age, ontogenetic age and experience.
Brain structure & function (2018-05-26)
S M Ohline, K L Wake, M-V Hawkridge, M F Dinnunhan, R U Hegemann, A Wilson, L Schoderboeck, B J Logan, T Jungenitz, S W Schwarzacher, S M Hughes, W C Abraham
ABSTRACT
Early during their maturation, adult-born dentate granule cells (aDGCs) are particularly excitable, but eventually develop the electrophysiologically quiet properties of mature cells. However, the stability versus plasticity of this quiet state across time and experience remains unresolved. By birthdating two populations of aDGCs across different animal ages, we found for 10-month-old rats the expected reduction in excitability across cells aged 4-12 weeks, as determined by Egr1 immunoreactivity. Unexpectedly, cells 35 weeks old (after genesis at an animal age of 2 months) were as excitable as 4-week-old cells, in the dorsal hippocampus. This high level of excitability at maturity was specific for cells born in animals 2 months of age, as cells born later in life did not show this effect. Importantly, excitability states were not fixed once maturity was gained, but were enhanced by enriched environment exposure or LTP induction, indicating that any maturational decrease in excitability can be compensated by experience. These data reveal the importance of the animal's age for aDGC excitability, and emphasize their prolonged capability for plasticity during adulthood.
Sigma-Aldrich
5-Chloro-2′-deoxyuridine, thymidine analog