Merck
Regulation of energy rheostasis by the melanocortin-3 receptor.
Science advances (2018-08-25)
Masoud Ghamari-Langroudi, Isin Cakir, Rachel N Lippert, Patrick Sweeney, Michael J Litt, Kate L J Ellacott, Roger D Cone
ABSTRACT
Like most homeostatic systems, adiposity in mammals is defended between upper and lower boundary conditions. While leptin and melanocortin-4 receptor (MC4R) signaling are required for defending energy set point, mechanisms controlling upper and lower homeostatic boundaries are less well understood. In contrast to the MC4R, deletion of the MC3R does not produce measurable hyperphagia or hypometabolism under normal conditions. However, we demonstrate that MC3R is required bidirectionally for controlling responses to external homeostatic challenges, such as caloric restriction or calorie-rich diet. MC3R is also required for regulated excursion from set point, or rheostasis, during pregnancy. Further, we demonstrate a molecular mechanism: MC3R provides regulatory inputs to melanocortin signaling, acting presynaptically on agouti-related protein neurons to regulate γ-aminobutyric acid release onto anorexigenic MC4R neurons, exerting boundary control on the activity of MC4R neurons. Thus, the MC3R is a critical regulator of boundary controls on melanocortin signaling, providing rheostatic control on energy storage.
Sigma-Aldrich
LY2112688 trifluoroacetate, ≥95% (HPLC)