Merck
  • Fusion of FIG to the receptor tyrosine kinase ROS in a glioblastoma with an interstitial del(6)(q21q21).
Fusion of FIG to the receptor tyrosine kinase ROS in a glioblastoma with an interstitial del(6)(q21q21).
Genes, chromosomes & cancer (2003-03-28)
Alain Charest, Keara Lane, Kevin McMahon, Julie Park, Elizabeth Preisinger, Helen Conroy, David Housman
ABSTRACT
The transmembrane proto-oncogene receptor tyrosine kinase (RTK) ROS is an orphan receptor that is aberrantly expressed in neoplasms of the central nervous system. Here, we report the fusion of its carboxy-terminal kinase domain to the amino-terminal portion of a protein called FIG (Fused in Glioblastoma) in a human glioblastoma multiforme (GBM). By characterizing both FIG and ROS genes in normal and in U118MG GBM cells, we determined that an intra-chromosomal homozygous deletion of 240 kilobases on 6q21 is responsible for the formation of the FIG-ROS locus. The FIG-ROS transcript is encoded by 7 FIG exons and 9 ROS-derived exons. We also demonstrate that the FIG-ROS locus encodes for an in-frame fusion protein with a constitutively active kinase activity, suggesting that FIG-ROS may act as an oncogene. This is the first example of a fusion RTK protein that results from an intra-chromosomal deletion, and it represents the first fusion RTK protein isolated from a human astrocytoma.
Sigma-Aldrich
ROS1 (1883-end), active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution