MilliporeSigma
  • Mice deficient for all PIM kinases display reduced body size and impaired responses to hematopoietic growth factors.
Mice deficient for all PIM kinases display reduced body size and impaired responses to hematopoietic growth factors.
Molecular and cellular biology (2004-06-17)
Harald Mikkers, Martijn Nawijn, John Allen, Conny Brouwers, Els Verhoeven, Jos Jonkers, Anton Berns
ABSTRACT
The Pim family of proto-oncogenes encodes a distinct class of serine/threonine kinases consisting of PIM1, PIM2, and PIM3. Although the Pim genes are evolutionarily highly conserved, the contribution of PIM proteins to mammalian development is unclear. PIM1-deficient mice were previously described but showed only minor phenotypic aberrations. To assess the role of PIM proteins in mammalian physiology, compound Pim knockout mice were generated. Mice lacking expression of Pim1, Pim2, and Pim3 are viable and fertile. However, PIM-deficient mice show a profound reduction in body size at birth and throughout postnatal life. In addition, the in vitro response of distinct hematopoietic cell populations to growth factors is severely impaired. In particular, PIM proteins are required for the efficient proliferation of peripheral T lymphocytes mediated by synergistic T-cell receptor and interleukin-2 signaling. These results indicate that members of the PIM family of proteins are important but dispensable factors for growth factor signaling.
Sigma-Aldrich
PIM3 active, GST tagged from mouse, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution