Merck
  • Production of chlorflavonin, an antifungal metabolite of Aspergillus candidus.
Production of chlorflavonin, an antifungal metabolite of Aspergillus candidus.
Applied microbiology (1970-05-01)
J E Munden, D Butterworth, G Hanscomb, M S Verrall
ABSTRACT
Production of chlorflavonin, a new antifungal antibiotic, by strains of Aspergillus candidus is described. Two wild strains of the fungus had distinctly different chlorflavonin-producing capabilities. One strain produced 25 mug of chlorflavonin per ml per 4 to 5 days in a pilot scale fermentor with stirring, using a medium containing corn steep liquor and glucose. Production of antibiotic was favored by high rates of agitation-aeration. Crude chlorflavonin was extracted from the whole brew with a hydrocarbon solvent and then purified by recrystallization from benzene and petroleum ether. The overall yield from fermentation brew to pure product was 50%.
Sigma-Aldrich
Chlorflavonin, ≥98% (HPLC)