Merck
  • Visible light-driven photocatalytic generation of sulfonamidyl radicals for alkene hydroamination of unsaturated sulfonamides.
Visible light-driven photocatalytic generation of sulfonamidyl radicals for alkene hydroamination of unsaturated sulfonamides.
Chemical communications (Cambridge, England) (2018-02-06)
Jun Chen, Hong-Mei Guo, Quan-Qing Zhao, Jia-Rong Chen, Wen-Jing Xiao
ABSTRACT
A visible light-driven photocatalytic generation of sulfonamidyl radicals, and application to intramolecular alkene hydroamination, has been accomplished, providing a mild and efficient approach to various functionalized isoxazolidines. The success of this protocol is based on the strategy of oxidative deprotonation electron transfer by merging the base and the photocatalyst under visible light irradiation, obviating installation of a photolabile handle or stoichiometric external oxidants.
Sigma-Aldrich
[Ru(phen)3]Cl2, ≥95%