Merck
  • Application of Fungicides and Microalgal Phenolic Extracts for the Direct Control of Fumonisin Contamination in Maize.
Application of Fungicides and Microalgal Phenolic Extracts for the Direct Control of Fumonisin Contamination in Maize.
Journal of agricultural and food chemistry (2018-04-28)
Priscila Tessmer Scaglioni, Massimo Blandino, Valentina Scarpino, Debora Giordano, Giulio Testa, Eliana Badiale-Furlong
ABSTRACT
Fungicides and, for the first time, microalgal phenolic extracts (MPE) from Spirulina sp. and Nannochloropsis sp. were applied on maize culture media under field conditions to evaluate their ability to minimize Fusarium species development and fumonisin production. An in vitro assay against F. verticillioides was carried out using maize grains as the culture medium. An open-field experiment was carried out in Northwest Italy under natural infection conditions. The compared treatments were factorial combinations of two insecticide treatments (an untreated control and pyrethroid, used against European Corn Borer), four antifungal treatments (an untreated control, MPE from Spirulina sp., MPE from Nannochloropsis sp., and a synthetic fungicide), and two timings of the application of the antifungal compounds (at maize flowering and at the milk stage). The MPE compounds were capable of inhibiting fumonisin production in vitro more efficiently than tebuconazole. Insecticide application reduced the infection by Fusarium species and subsequent fumonisin contamination. However, fumonisins in maize fields were not significantly controlled by either fungicide or MPE application.