Merck
  • Acaricide-Mediated Competition Between the Sibling Species Tetranychus cinnabarinus and Tetranychus urticae.
Acaricide-Mediated Competition Between the Sibling Species Tetranychus cinnabarinus and Tetranychus urticae.
Journal of economic entomology (2018-03-01)
Wencai Lu, Yuan Hu, Peng Wei, Qiang Xu, Christian Bowman, Ming Li, Lin He
ABSTRACT
The carmine spider mite (Tetranychus cinnabarinus [Acarifonnes: Tetranychidae]) and the two-spotted spider mite (Tetranychus urticae [Acarifonnes: Tetranychidae]) are two notorious pests of agricultural crops. Control of these pests has been dependent upon using different kinds of acaricides. The purpose of this study was to determine the differential responses of these two pest species collected from crops in the same field to acaricide treatments. Field trials have shown that without spraying acaricides, T. cinnabarinus will displace T. urticae. However, the application of abamectin has the potential to change the composition of spider mite complexes and facilitate the interspecific competition of T. urticae against T. cinnabarinus when both are fed on cowpeas and eggplants. Moreover, T. urticae is more prone to develop resistance than T. cinnabarinus when selected in the laboratory using cyflumetofen or fenpropathrin. After 20 generations of acaricide selection, the activities of detoxifying enzymes were considerably higher in T. urticae with more detoxifying enzymes upregulated after selection in this species. The results of this study demonstrate that differential responses to acaricide treatments have made it possible for T. urticae to overcome the competitive advantage present in T. cinnabarinus during the absence of acaricide application.
Product Description
Supelco
Cyflumetofen, PESTANAL®, analytical standard