Merck
  • Rapid Trace Detection and Isomer Quantitation of Pesticide Residues via Matrix-Assisted Laser Desorption/Ionization Fourier Transform Ion Cyclotron Resonance Mass Spectrometry.
Rapid Trace Detection and Isomer Quantitation of Pesticide Residues via Matrix-Assisted Laser Desorption/Ionization Fourier Transform Ion Cyclotron Resonance Mass Spectrometry.
Journal of agricultural and food chemistry (2018-03-29)
Xinzhou Wu, Weifeng Li, Pengran Guo, Zhixiang Zhang, Hanhong Xu
ABSTRACT
Matrix-assisted laser desorption/ionization Fourier transform ion cyclotron resonance mass spectrometry (MALDI-FTICR-MS) has been applied for rapid, sensitive, undisputed, and quantitative detection of pesticide residues on fresh leaves with little sample pretreatment. Various pesticides (insecticides, bactericides, herbicides, and acaricides) are detected directly in the complex matrix with excellent limits of detection down to 4 μg/L. FTICR-MS could unambiguously identify pesticides with tiny mass differences (∼0.017 75 Da), thereby avoiding false-positive results. Remarkably, pesticide isomers can be totally discriminated by use of diagnostic fragments, and quantitative analysis of pesticide isomers is demonstrated. The present results expand the horizons of the MALDI-FTICR-MS platform in the reliable determination of pesticides, with integrated advantages of ultrahigh mass resolution and accuracy. This method provides growing evidence for the resultant detrimental effects of pesticides, expediting the identification and evaluation of innovative pesticides.
Product Description
Supelco
Oxasulfuron, PESTANAL®, analytical standard