Skip to Content
Merck
Z-VAD-FMK solid

Z-VAD-FMK

Z-VAD-FMK solid
Synonym(s):
N-Benzyloxycarbonyl-Val-Ala-Asp(O-Me) fluoromethyl ketone
Empirical Formula (Hill Notation):
C22H30FN3O7
CAS Number:
Molecular Weight:
467.49

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing