Skip to Content
Merck
Xanthohumol primary reference standard

Xanthohumol

Xanthohumol primary reference standard
Synonym(s):
(E)-1-[2,4-Dihydroxy-6-methoxy-3-(3-methyl-2-butenyl)phenyl]-3-(4-hydroxyphenyl)propenone, 2′,4,4′-Trihydroxy-6′-methoxy-3′-prenylchalcone
Empirical Formula (Hill Notation):
C21H22O5
CAS Number:
Molecular Weight:
354.40

Products

Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing