![Val-cit-PAB-OH ≥95%](/deepweb/assets/sigmaaldrich/product/structures/183/763/6f05e4e0-a056-4a55-b837-584167d0b5b2/640/6f05e4e0-a056-4a55-b837-584167d0b5b2.png)
Val-cit-PAB-OH
![Val-cit-PAB-OH ≥95%](/deepweb/assets/sigmaaldrich/product/structures/183/763/6f05e4e0-a056-4a55-b837-584167d0b5b2/640/6f05e4e0-a056-4a55-b837-584167d0b5b2.png)
Synonym(s):
L-Valyl-N5-(aminocarbonyl)-N-[4-(hydroxymethylphenyl]-L-ornithinamide
Empirical Formula (Hill Notation):
C18H29N5O4
CAS Number:
Molecular Weight:
379.45
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing