Skip to Content
Merck
Tris(phenylthio)methane 98%

Tris(phenylthio)methane

Tris(phenylthio)methane 98%
Synonym(s):
Triphenyl trithioorthoformate
Linear Formula:
(C6H5S)3CH
CAS Number:
Molecular Weight:
340.53

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing