Skip to Content
Merck
Tris[2-(p-tolyl)pyridine]iridium(III) ≥99% (HPLC)

Tris[2-(p-tolyl)pyridine]iridium(III)

Tris[2-(p-tolyl)pyridine]iridium(III) ≥99% (HPLC)
Synonym(s):
Ir(p-Me-ppy)3
Empirical Formula (Hill Notation):
C36H30IrN3
CAS Number:
Molecular Weight:
696.86

Products

Brand
Formula Weight
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing