
Trazodone for system suitability

Synonym(s):
Trazodone hydrochloride, 2-[3-[4-(3-Chlorophenyl)-1-piperazinyl]propyl]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one hydrochloride
Empirical Formula (Hill Notation):
C19H22ClN5O · HCl
CAS Number:
Molecular Weight:
408.32
Products
Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing