![TOYOPEARL GigaCap® CM-650M MiniChrom Column phase carboxymethyl, 75 μm particle size, pore size 1000 Å, L × I.D. 10 cm × 8 mm, MiniChrom Column](/deepweb/assets/sigmaaldrich/product/images/374/600/7f529074-0503-4e9e-8945-8a0cf4551927/640/7f529074-0503-4e9e-8945-8a0cf4551927.jpg)
TOYOPEARL GigaCap® CM-650M MiniChrom Column
![TOYOPEARL GigaCap® CM-650M MiniChrom Column phase carboxymethyl, 75 μm particle size, pore size 1000 Å, L × I.D. 10 cm × 8 mm, MiniChrom Column](/deepweb/assets/sigmaaldrich/product/images/374/600/7f529074-0503-4e9e-8945-8a0cf4551927/640/7f529074-0503-4e9e-8945-8a0cf4551927.jpg)
Synonym(s):
MiniChrom
Products
Product Category
Manufacturer
Product Line
Analyte Suitability
Separation Technique
ID (mm)
Length (cm)
Volume (ml)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing