[Ru(bpm)3][Cl]2
Empirical Formula (Hill Notation):
C24H18Cl2N12Ru
CAS Number:
Molecular Weight:
646.46
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing