Skip to Content
MX
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
Spare sealing disc for high temperature screwcaps
Products
Manufacturer
Color
Special Grade
Natural (3)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
SLW4546/32
Pyrex
®
Spare sealing disc for high temperature screwcaps,
natural, GL 32
Expand
View Pricing
SLW4546/25
Pyrex
®
Spare sealing disc for high temperature screwcaps,
natural, GL 25
Expand
View Pricing
SLW4546/45
Pyrex
®
Spare sealing disc for high temperature screwcaps,
natural, GL 45
Expand
View Pricing