![Propyphenazone European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/181/780/905e9f24-aa9e-4939-a048-17ca092206f7/640/905e9f24-aa9e-4939-a048-17ca092206f7.png)
Propyphenazone
![Propyphenazone European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/181/780/905e9f24-aa9e-4939-a048-17ca092206f7/640/905e9f24-aa9e-4939-a048-17ca092206f7.png)
Synonym(s):
4-Isopropylantipyrine, 1,2-Dihydro-1,5-dimethyl-4-(1-methylethyl)-2-phenyl-3H-pyrazol-3-one, 1,2-Dihydro-4-isopropyl-1,5-dimethyl-2-phenyl-3H-pyrazol-3-one, Isopropylphenazone, Propyphenazone
Empirical Formula (Hill Notation):
C14H18N2O
CAS Number:
Molecular Weight:
230.31
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing