Skip to Content
Merck
Poly(ethylene adipate) average Mw ~10,000 by GPC

Poly(ethylene adipate)

Poly(ethylene adipate) average Mw ~10,000 by GPC
Synonym(s):
Ethylene glycol adipate, Poly(ethylene adipate), dihydroxy terminated
Linear Formula:
[OCH2CH2O2C(CH2)4CO]n
CAS Number:

Products

Brand
Physical Form
Polymer Composition
Polymer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing