![Phosphoglucomutase from rabbit muscle](/deepweb/assets/sigmaaldrich/product/images/339/852/1cc86871-52ff-4de4-bde9-c78f4ebbcc4d/640/1cc86871-52ff-4de4-bde9-c78f4ebbcc4d.jpg)
Phosphoglucomutase from rabbit muscle
![Phosphoglucomutase from rabbit muscle](/deepweb/assets/sigmaaldrich/product/images/339/852/1cc86871-52ff-4de4-bde9-c78f4ebbcc4d/640/1cc86871-52ff-4de4-bde9-c78f4ebbcc4d.jpg)
Synonym(s):
α-D-Glucose-1,6-bisphosphatase, α-D-Glucose-1-phosphate phosphotransferase
CAS Number:
Products
Brand
Biological Source
Color
Physical Form
Tissue
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing