Skip to Content
Merck
Phenethyl acetate analytical standard

Phenethyl acetate

Phenethyl acetate analytical standard
Synonym(s):
2-Phenethyl acetate, 2-Phenylethyl acetate
Linear Formula:
CH3COOCH2CH2C6H5
CAS Number:
Molecular Weight:
164.20
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing