Skip to Content
Merck
Perphenazine Related Compound B certified reference material, pharmaceutical secondary standard

Perphenazine Related Compound B

Perphenazine Related Compound B certified reference material, pharmaceutical secondary standard
Synonym(s):
2-{4-[3-(10H-Phenothiazin-10-yl)propyl]piperazin-1-yl}ethanol
Empirical Formula (Hill Notation):
C21H27N3OS
CAS Number:
Molecular Weight:
369.52

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade