![Pazufloxacin VETRANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/271/095/db878ede-60d9-4bb9-ab7f-e49a91dd3afc/640/db878ede-60d9-4bb9-ab7f-e49a91dd3afc.png)
Pazufloxacin
![Pazufloxacin VETRANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/271/095/db878ede-60d9-4bb9-ab7f-e49a91dd3afc/640/db878ede-60d9-4bb9-ab7f-e49a91dd3afc.png)
Empirical Formula (Hill Notation):
C16H15FN2O4
CAS Number:
Molecular Weight:
318.30
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing